missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Monoclonal Antibody (MEM-263), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110228
218.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD44 Monoclonal antibody specifically detects CD44 in Canine, Human, Non-human Primate, Porcine samples. It is validated for Flow Cytometry.Specifications
CD44 | |
Monoclonal | |
FITC | |
CD44 | |
AU023126; AW121933; AW146109; Bovine CD44; CD44; CD44 antigen; CD44 antigen-like protein; CD44 molecule; CD44 molecule (Indian blood group); CD44 protein; CD44 variant; CD44A; CD44-like protein; CDW44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; extracellular matrix receptor-III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IN; LHR; Ly-24; lymphocyte antigen 24; lymphocyte surface antigen precursor CD44; MC56; MDU2; MDU3; METAA; MIC4; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; phagocytic glycoprotein I; putative Bos taurus CD44; RHAMM; sCD 44; sCD44; soluble CD 44; soluble CD44 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
100525150, 103237736, 403939, 960 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-263 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P16070, Q28284 | |
CD44 | |
COS-7 cells (African Green Monkey). | |
100 Tests | |
Primary | |
Canine, Human, Non-human Primate, Porcine | |
Antibody | |
IgG1 |