Learn More
CD42b Mouse anti-Human, Unlabeled, Clone: HIP1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555471
Additional Details : Weight : 0.01000kg
Description
The HIP1 monoclonal antibody specifically binds to CD42b. CD42b is also known as the Platelet glycoprotein Ib alpha chain that is encoded by the GP1BA gene. CD42b is disulfide bonded to CD42c to form a 170 kDa heterodimer, GPIb. GPIb forms a noncovalent complex with CD42a and CD42d (CD42 complex) that is expressed on platelets and megakaryocytes. The CD42 complex serves as the von Willebrand Factor(vWF) surface receptor involved in the adhesion of platelets to the subendothelium of damaged vascular walls. HIP1 inhibits the ristocetin-dependent binding ofvWF to platelets and partially inhibits collagen-induced aggregation.
Specifications
CD42b | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
HIP1 | |
Unconjugated | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
For Research Use Only.