Learn More
CD42a Mouse anti-Human, Clone: GR-P, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191023
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-91023 detects CD42a from human samples. MA1-91023 has been successfully used in immunoprecipitation applications. The MA1-91023 immunogen is: Human red blood cells and platelets Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, wSpecifications
CD42a | |
Monoclonal | |
Unconjugated | |
P14770 | |
GP9 | |
Human red blood cells and platelets | |
200 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
GR-P | |
GP9 | |
CD42a, GPIX, GP-IX, glycoprotein 9, platelet glycoprotein IX, Platelet GPIX | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
2815 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |