missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD154 (CD40 Ligand) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA126457
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-26457 detects CD40L from human samples.
CD154 (CD40 Ligand) Polyclonal antibody specifically detects CD154 (CD40 Ligand) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
CD154 (CD40 Ligand) | |
Polyclonal | |
Unconjugated | |
CD40LG | |
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21947, 84349, 959 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.5% BSA, 30% glycerol, 5mM EDTA and 0.03% ProClin 300; pH 7.2 | |
P27548, P29965, Q9Z2V2 | |
CD40LG | |
Synthetic peptide surrounding amino acid 246 of human CD40 Ligand | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |