Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (40804), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523829
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CD27 Ligand, rhCD30 Ligand, or recombinant mouse CD40 Ligand is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CD154 (CD40 Ligand) Monoclonal antibody specifically detects CD154 (CD40 Ligand) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot.Specifications
CD154 (CD40 Ligand) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P29965 | |
CD40LG | |
E. coli-derived recombinant human CD40 Ligand/TNFSF5 Leu50-Leu261 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot | |
40804 | |
Unconjugated | |
CD40LG | |
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
Mouse | |
Protein A/G | |
RUO | |
959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |