Learn More
CD40 Monoclonal Antibody (3/23), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA181395
354.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse CD40 antibody, clone 3/23 recognizes the murine CD40 cell surface glycoprotein.
CD40 Monoclonal antibody specifically detects CD40 in Mouse samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen).Specifications
CD40 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P27512 | |
CD40 | |
Extracellular domain of Murine CD40 and the Fc portion of Human IgG1. | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
3/23 | |
Unconjugated | |
CD40 | |
AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5 | |
Rat | |
Protein G | |
RUO | |
21939 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |