missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse anti-Human, Clone: 34915, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3791

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460256

  • £337.93 / 500µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

CD4 Monoclonal antibody MAB3791 Specifically detects CD4 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CD4
34915
Western Blot 1 ug/mL
P01730
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human CD4 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse CD4.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD4
Recombinant human CD4, Extracellular domain, Accession # P01730
Protein A or G purified from ascites
RUO
Primary
920
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only