Learn More
CD4 Rat anti-Mouse, APC-H7, Clone: GK1.5, BD
Rat Monoclonal Antibody
£149.00 - £295.00
Specifications
Antigen | CD4 |
---|---|
Clone | GK1.5 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15820049
|
BD Biosciences
560246 |
25 μg |
£149.00
Each |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899988
|
BD Biosciences
560181 |
0.1 mg |
£295.00
Each |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The GK1.5 clone has been reported to react with the CD4 (L3T4) differentiation antigen expressed on most thymocytes, a subpopulation of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells), and a subset of NK-T cells. In addition, CD4 has also been reported to be detectable on pluripotent hematopoietic stem cells, bone marrow myeliod and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 has also been reported to be expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. GK1.5 mAb reportedly blocks binding of the RM4-5 (Mfr. No. 553046/553047) and H129.19 (Mfr. No. 553650/553651), but not RM4-4 (Mfr. No. 553055) antibodies.
Host Species: Rat
Clone: GK1.5
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse CTL clone V4
Flow Cytometry, Intracellular Staining
Specifications
CD4 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 |
GK1.5 | |
Flow Cytometry | |
APC-H7 | |
RUO | |
Mouse | |
Mouse CTL clone V4 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.