missing translation for 'onlineSavingsMsg'
Learn More

CD36/SR-B3 Rat anti-Human, Clone: 255619, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1955

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469055

  • £311.64 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD36/SR-B3 Monoclonal antibody MAB1955 Specifically detects CD36/SR-B3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CD36/SR-B3
255619
Western Blot 1 ug/mL
P16671
CD36, CD36 antigen, CD36 molecule (thrombospondin receptor), Collagen R, FA6-152, FAT, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIb, GPIV, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, SCARB3, scavenger receptor class B, member 3, SRB3, SR-B3, Thrombospondin R, Thrombospondin receptor
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CD36 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) CD36 is observed. In Western blots, no cross-reactivity with rmCD36 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD36
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD36/SR-B3 Gly30-Asn439 Accession # P16671
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
948
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only