Learn More
CD35 Monoclonal Antibody (To5), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517715
143.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17715 targets CD35/CR1 in FACS applications and shows reactivity with Human samples. The MA5-17715 immunogen is mouse monoclonal antibodies raised against human C3b receptor-enriched precipitates from tonsil cell membranes.
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Flow Cytometry.Specifications
CD35 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P17927 | |
CR1 | |
Mouse monoclonal antibodies raised against human C3b receptor-enriched precipitates from tonsil cell membranes. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
To5 | |
PE | |
CR1 | |
C3 binding protein; C3b/C4b receptor; C3-binding protein; C3BR; C4BR; CD35; CD35 antigen; complement component (3b/4b) receptor 1 (Knops blood group); Complement Component (3b/4b) receptor 1 including Knops blood group system; complement component 3b/4b receptor 1 (Knops blood group); complement receptor type 1; CR1; KN; Knops blood group antigen | |
Mouse | |
Protein G | |
RUO | |
1378 | |
4° C, do not freeze | |
Liquid |