Learn More
ErbB2 (HER-2) Monoclonal Antibody (ICR55), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA182367
Code : LD
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Human HER2/neu antibody, clone ICR55 recognizes epitope E of human HER2/neu, otherwise known as CD340 and c-ErbB-2, a proto-oncogene expressed at the cell surface of a range of tumor cells, often associated with poor prognosis, and an increased risk of metastasis.
ErbB2 (HER-2) Monoclonal antibody specifically detects ErbB2 (HER-2) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
ErbB2 (HER-2) | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P04626 | |
ERBB2 | |
Proprietary Immunogen. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
ICR55 | |
Unconjugated | |
ERBB2 | |
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb | |
Rat | |
Protein G | |
RUO | |
2064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |