Learn More
CD34 Monoclonal Antibody (MEC14.7), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA122646
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody detects a neuraminidase sensitive epitope on endothelium in vivo, particullary on small vessels and neoformed capillaries and develpoing vascular structures in embryonal structures. Furthermore, the antibody reacts with hematopoietic progenitors particularly of myelomonocytic colony forming cells.
CD34 Monoclonal antibody specifically detects CD34 in Human, Mouse, Rabbit samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD34 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P28906, Q64314 | |
Cd34 | |
Mouse CD34 | |
100 μg | |
Primary | |
Human, Mouse, Rabbit | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
MEC14.7 | |
Unconjugated | |
Cd34 | |
(E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 | |
Rat | |
Protein G | |
RUO | |
100343670, 12490, 947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |