Learn More
CD34 Monoclonal Antibody (QBEND/10), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516925
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody does not react with bovine, dog, rat or sheep.|For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.|Mouse anti Human CD34 antibody, clone QBEND/10 recognizes the human CD34 antigen, also known as Hematopoietic progenitor cell antigen CD34.|Quantity: 100 Tests/1 mL
CD34 Monoclonal antibody specifically detects CD34 in Cynomolgus Monkey, Human samples. It is validated for Flow Cytometry.Specifications
CD34 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P28906 | |
Cd34 | |
Human endothelial cell membrane vesicles | |
100 Tests | |
Primary | |
Cynomolgus Monkey, Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
QBEND/10 | |
FITC | |
Cd34 | |
(E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 | |
Mouse | |
purified | |
RUO | |
101866926, 947 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |