missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Monoclonal Antibody (FOS), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112100
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow Cytometry.Specifications
CD33 | |
Monoclonal | |
30 μg/mL | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P20138 | |
CD33 | |
FMY9S5 cells containing the CD33 gene | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
FOS | |
PE | |
CD33 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
Mouse | |
Protein A/G | |
RUO | |
945 | |
4° C, do not freeze | |
Liquid |