Learn More
CD33 Monoclonal Antibody (2B7C12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515446
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15446 targets CD33 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15446 immunogen is purified recombinant fragment of CD33 (48-258) expressed in E. Coli.
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for ELISA, Western Blot.Specifications
CD33 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P20138 | |
CD33 | |
Purified recombinant fragment of CD33 (48-258) expressed in E. Coli. | |
RUO | |
945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
2B7C12 | |
Unconjugated | |
CD33 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |