Learn More
EpCAM Monoclonal Antibody (VU-1D9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516973
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. A suggested positive control for immunohistochemical applications is large bowel. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. This product is a low-endotoxin, preservative-free formulation. Mouse anti Human CD326 antibody, clone VU-1D9 recognizes epithelial cell adhesion molecule (Ep-CAM), a approximately 34 kDa cell surface antigen otherwise known as CD326, MOC31 or Ber-EP4.
EpCAM Monoclonal antibody specifically detects EpCAM in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
EpCAM | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P16422 | |
EPCAM | |
HG9 cell line (small cell lung carcinoma) | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
VU-1D9 | |
Unconjugated | |
EPCAM | |
adenocarcinoma-associated antigen; CD326; cell surface glycoprotein Trop-1; DIAR5; EGP; EGP-2; EGP314; EGP40; EPCAM; Ep-CAM; EpCAM1; epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; GA733-2; gp40; hEGP314; HNPCC8; human epithelial glycoprotein-2; KS 1/4 antigen; KS1/4; KSA; Ly74; lymphocyte antigen 74; M1S2; M4S1; major gastrointestinal tumor-associated protein GA733-2; mEGP314; membrane component, chromosome 4, surface marker (35kD glycoprotein); MIC18; MK-1; panepithelial glycoprotein 314; protein 289A; Protein D5.7A; Tacsd1; Tacstd1; TROP1; Trop-1 protein; Tumor-associated calcium signal transducer 1 | |
Mouse | |
Protein A | |
RUO | |
4072 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |