Learn More
E-cadherin Monoclonal Antibody (67A4), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA110192
Code : LD
Additional Details : Weight : 0.01000kg
Product Code. 12352213
  • £348.00 / Pack of 100
Estimated Shipment: 08-05-2024
to see stock.
Description
Description
This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. [provided by RefSeq, Jul 2008]
E-cadherin Monoclonal antibody specifically detects E-cadherin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.
Specifications
Specifications
E-cadherin
Monoclonal
1.0 mg/mL
PBS with 15mM sodium azide; pH 7.4
P12830
CDH1
T-47D cells
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
67A4
Unconjugated
CDH1
AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin
Mouse
Protein A
RUO
999
4° C, do not freeze