missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-cadherin Monoclonal Antibody (67A4), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110193
Code : LD
Additional Details : Weight : 0.01000kg
Description
E-cadherin Monoclonal antibody specifically detects E-cadherin in Human samples. It is validated for Flow Cytometry.Specifications
E-cadherin | |
Monoclonal | |
APC | |
CDH1 | |
AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
999 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
67A4 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P12830 | |
CDH1 | |
T-47D cells | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |