Learn More
CD31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516301
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: IHC (P): Mouse skin and xenograft tissue, human tonsil tissue WB: Jurkat whole cell lysate, human vascular endothelial cell lysate IHC (F): Human fetal heart tissue
CD31 Polyclonal antibody specifically detects CD31 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD31 | |
Polyclonal | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5175 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.013 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P16284 | |
Pecam1 | |
Synthetic peptide corresponding to mouse CD31, aa 700 to the C-terminus. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |