missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529166
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HUVEC. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD31 Polyclonal antibody specifically detects CD31 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD31 | |
Polyclonal | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P16284 | |
Pecam1 | |
Recombinant fragment corresponding to a region within amino acids 288 and 552 of Human CD31 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |