missing translation for 'onlineSavingsMsg'
Learn More

CD31/PECAM-1 Rat anti-Porcine, Clone: 377521, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3387

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460046

  • £249.49 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

CD31/PECAM-1 Monoclonal antibody MAB3387 Specifically detects CD31/PECAM-1 in Porcine samples. It is validated for Western Blotting
Specifications

Specifications

CD31/PECAM-1
377521
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PECAM1
Mouse myeloma cell line NS0-derived recombinant porcine CD31/PECAM-1, Gln28-Lys602 (predicted), Accession # Q95242
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5175
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q95242
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects porcine CD31/PECAM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CD31, rhCECAM-1, rhICAM-1, rhICAM-2, rhICAM-3, rhVCAM-1, recombinant mouse (rm) MadCAM-1, rmCD31, or rmICAM-5 is observed.
Porcine
Videos

For Research Use Only