Learn More
CD31 Monoclonal Antibody (TLD-3A12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516951
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD31 Monoclonal antibody specifically detects CD31 in Human, Rat, Rhesus Monkey samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Specifications
CD31 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P16284, Q3SWT0 | |
Pecam1 | |
Activated, Lewis rat derived microglial cells | |
500 μg | |
Primary | |
Human, Rat, Rhesus Monkey | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
TLD-3A12 | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
Protein G | |
RUO | |
29583, 5175, 718302 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |