Learn More
CD31 Monoclonal Antibody (SP38), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516337
Code : PN
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil tissue.
CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CD31 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant diluted in TBS with 1% BSA and 0.1% sodium azide; pH 7.5 | |
P16284 | |
Pecam1 | |
Synthetic peptide corresponding to C-terminus of mouse CD31 | |
RUO | |
5175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
SP38 | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Rabbit | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |