Learn More
CD31 Monoclonal Antibody (LCI-4), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516770
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Pig CD31, clone LCI-4 recognizes porcine CD31, also known as Platelet endothelial cell adhesion molecule (PECAM-1).
CD31 Monoclonal antibody specifically detects CD31 in Human, Porcine samples. It is validated for Flow Cytometry.Specifications
CD31 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P16284, Q95242 | |
Pecam1 | |
Porcine CD31/human IgGFc fusion protein | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Flow Cytometry | |
LCI-4 | |
Biotin | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
Protein A | |
RUO | |
396941, 5175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |