Learn More
CD153 Monoclonal Antibody (116614), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523859
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) CD27 Ligand, recombinant mouse CD30 Ligand, or rhCD40 Ligand is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CD153 Monoclonal antibody specifically detects CD153 in Human samples. It is validated for Flow Cytometry, Neutralization.Specifications
CD153 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P32971 | |
Tnfsf8 | |
Mouse myeloma cell line NS0-derived recombinant human CD30 Ligand/TNFSF8 Gln63-Asp234 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Neutralization | |
116614 | |
Unconjugated | |
Tnfsf8 | |
CD153; CD153 antigen; CD30 antigen ligand; CD30 ligand; Cd30l; CD30-L; Cd30lg; CD30ligand; Tnfsf8; TNLG3A; tumor necrosis factor (ligand) superfamily member 8; tumor necrosis factor (ligand) superfamily, member 8; tumor necrosis factor ligand 3A; tumor necrosis factor ligand superfamily member 8; tumor necrosis factor superfamily member 8 | |
Mouse | |
Protein A/G | |
RUO | |
944 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |