Learn More
CD300f/LMIR3 Rat anti-Mouse, Clone: 305835, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB27881
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439695
  • £271.93 / 100µg
Estimated Shipment
27-06-2019
Description
Description
CD300f/LMIR3 Monoclonal antibody MAB27881 Specifically detects CD300f/LMIR3 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
CD300f/LMIR3/CD300LF
305835
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD300LF
Mouse myeloma cell line NS0-derived recombinant mouse CD300f/LMIR3 Cys16-Gly188 Accession # Q6SJQ7
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
146722
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Q6SJQ7
CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CD300LF, CLIM1, CLM1, CLM-1, DIgR2, IgSF13, IREM1, IREM-1, LMIR3, NKIR, Pigr3
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse CD300f/LMIR3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) LMIR4, rmCD300b/LMIR5, recombinant human (rh) CD300a/LMIR1, rhCD300c/LMIR2, rhCD300f/LMIR3, or rhCD300e/LMIR6 is observed.