Learn More
CD30 Monoclonal Antibody (3B10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517044
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17044 targets CD30 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17044 immunogen is purified recombinant fragment of human CD30 expressed in E. Coli.
CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for ELISA, Flow Cytometry, Western Blot.Specifications
CD30 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P28908 | |
TNFRSF8 | |
Purified recombinant fragment of human CD30 expressed in E. Coli. | |
RUO | |
943 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Western Blot | |
3B10 | |
Unconjugated | |
TNFRSF8 | |
Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |