missing translation for 'onlineSavingsMsg'
Learn More

CD3, clone: OKT3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0037-82

Code : Z2

Additional Details : Weight : 0.01000kg

Product Code. 15276737

  • £83.11 / 100µg


EU Stock 13
Add to basket

Description

Description

Description: The OKT3 monoclonal antibody reacts with an epitope on the epsilon-subunit within the human CD3 complex. The OKT3 antibody has been reported to have potent immunosuppressive properties in vivo and has been proven effective in the treatment of renal, heart and liver allograft rejection. The CD3 subunits, gamma, delta, and epsilon chains, are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.Applications Reported: The OKT3 antibody has been reported for use in flow cytometric analysis. OKT3 has also been reported in in vitro activation of T cells. (Please use Functional Grade purified OKT3, cat. 16-0037, in functional assays.).Applications Tested: The OKT3 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD3
Monoclonal
0.5mg/mL
TBS with 0.09% sodium azide; pH 8.0
P04234, P07766, P09693, P20963
FLJ18683, T3E, TCRE
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
OKT3
Unconjugated
CD3E
Purified
CD247, CD3D, CD3E, CD3G
IgG2a, kappa
100μg
Store at 2-8°C.
Primary
915, 916, 917, 919
Videos

For Research Use Only.