Learn More
TLR10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529364
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLa, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (97%), Chimpanzee (100%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TLR10 Polyclonal antibody specifically detects TLR10 in Human samples. It is validated for Western Blot.Specifications
TLR10 | |
Polyclonal | |
Unconjugated | |
TLR10 | |
CD290; MGC104967; MGC126398; MGC126399; TLR10; toll like receptor 10; toll-like receptor 10; UNQ315/PRO358 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81793 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9BXR5 | |
TLR10 | |
Recombinant fragment corresponding to a region within amino acids 547 and 772 of Human CD290(TLR10) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |