missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGB1 Monoclonal Antibody (4B7R), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513658
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-13658 targets CD29 in FACS, IF, and IHC (F) applications and shows reactivity with Human and Porcine samples. The MA5-13658 immunogen is human melanoma V+B2 cell line.
ITGB1 Monoclonal antibody specifically detects ITGB1 in Human, Porcine samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen).Specifications
ITGB1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P05556, Q9GLP0 | |
ITGB1 | |
Human melanoma V+B2 cell line | |
500 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Frozen) | |
4B7R | |
Unconjugated | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
Protein G | |
RUO | |
3688, 397019 | |
4° C | |
Liquid |