Learn More
ITGB1 Monoclonal Antibody (12G10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180764
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody does not react with mouse or rat. Mouse anti Human CD29 monoclonal antibody, clone 12G10 recognizes human CD29 also known as beta1 integrin or VLA-4 subunit alpha.
ITGB1 Monoclonal antibody specifically detects ITGB1 in Human, Mink samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Specifications
ITGB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P05556 | |
ITGB1 | |
Purified human beta1 integrin preparation from HT1080 fibrosarcoma cell extract. | |
200 μg | |
Primary | |
Human, Mink | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
12G10 | |
Unconjugated | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
Protein G | |
RUO | |
3688 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |