missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Monoclonal Antibody (HTA125), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516719
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
TLR4 Monoclonal antibody specifically detects TLR4 in Canine, Guinea Pig, Human, Porcine, Rhesus Monkey samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
TLR4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
O00206, Q68Y56 | |
TLR4 | |
Ba/F3 cell line expressing TLR4 (CD284) | |
200 μg | |
Primary | |
Canine, Guinea Pig, Human, Porcine, Rhesus Monkey | |
Antibody | |
IgG2a |
Flow Cytometry, Immunoprecipitation | |
HTA125 | |
Unconjugated | |
TLR4 | |
ARMD10; CD284; EGK_07381; homolog of Drosophila toll; hToll; lipopolysaccharide response; Lps; Ly87; Ran/M1; Rasl2-8; Tlr4; TLR-4; TOLL; toll like receptor 4; toll/interleukin-l receptor (TIR) domain; toll4; toll-like receptor 4; toll-like receptor 4 immunity-related protein; Toll-like receptor 4-like protein; Toll-like receptor4 protein | |
Mouse | |
Protein G | |
RUO | |
100724803, 399541, 403417, 574360, 7099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |