Learn More
TLR4 Monoclonal Antibody (HTA125), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180122
Code : LD
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD284 antibody, clone HTA125 recognizes the human Toll like receptor 4 (TLR4) cell surface antigen.
TLR4 Monoclonal antibody specifically detects TLR4 in Canine, Guinea Pig, Human, Porcine, Rhesus Monkey samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation.Specifications
TLR4 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O00206, Q68Y56 | |
TLR4 | |
Ba/F3 cell line expressing TLR4 (CD284). | |
500 μg | |
Primary | |
Canine, Guinea Pig, Human, Porcine, Rhesus Monkey | |
Antibody | |
IgG2a |
Flow Cytometry, Functional Assay, Immunoprecipitation | |
HTA125 | |
Unconjugated | |
TLR4 | |
ARMD10; CD284; EGK_07381; homolog of Drosophila toll; hToll; lipopolysaccharide response; Lps; Ly87; Ran/M1; Rasl2-8; Tlr4; TLR-4; TOLL; toll like receptor 4; toll/interleukin-l receptor (TIR) domain; toll4; toll-like receptor 4; toll-like receptor 4 immunity-related protein; Toll-like receptor 4-like protein; Toll-like receptor4 protein | |
Mouse | |
Protein A | |
RUO | |
100724803, 399541, 403417, 574360, 7099 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |