missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Monoclonal Antibody (JJ319), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517472
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD28 Monoclonal antibody specifically detects CD28 in Rat samples. It is validated for Flow Cytometry.Specifications
CD28 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P31042 | |
Cd28 | |
Rat CD28 transfected A20J cells | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
JJ319 | |
FITC | |
Cd28 | |
antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
Mouse | |
Protein G | |
RUO | |
25660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |