Learn More
CD28 Monoclonal Antibody (B-23), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA112260
Code : LD
Additional Details : Weight : 0.01000kg
Product Code. 11578053
  • £363.00 / 100 tests
Estimated Shipment: 08-05-2024
to see stock.
Description
Description
CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary’s Disease.
Specifications
Specifications
CD28
Monoclonal
APC
P10747
CD28
HPB-ALL T-cell line
100 Tests
Primary
Human
Antibody
IgG1
Flow Cytometry
B-23
CD28
MGC138290, Tp44, CD28 antigen, T-cell-specific surface glycoprotein CD28
Mouse
Protein A/G
RUO
940
4° C, do not freeze