Learn More
PD-1 Monoclonal Antibody (J43), Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen PA535008
218.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PA5-35008 targets CD279/PD1 in FACS applications and shows reactivity with Mouse samples. The PA5-35008 immunogen is pD-1 cDNA transfectant.
PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
PD-1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q02242 | |
Pdcd1 | |
PD-1 cDNA transfectant | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
J43 | |
Unconjugated | |
Pdcd1 | |
CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
Armenian Hamster | |
Protein G | |
RUO | |
18566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |