Learn More
ICOS (CD278) Monoclonal Antibody (SP98), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516415
Code : PN
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil or reactive lymph node tissue.
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
ICOS (CD278) | |
Monoclonal | |
Conc. not determined | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
Q9Y6W8 | |
ICOS | |
Synthetic peptide corresponding to the C-terminus of human CD278 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
SP98 | |
Unconjugated | |
ICOS | |
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
Rabbit | |
Protein A/G | |
RUO | |
29851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |