Learn More
B7-H3 Monoclonal Antibody (6A1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515693
Code : A0
Detalles adicionales : Weight : 0.01000kg
Descripción
MA5-15693 targets CD276 in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human samples. The MA5-15693 immunogen is purified recombinant fragment of human CD276 expressed in E. Coli. MA5-15693 detects CD276 which has a predicted molecular weight of approximately 57kDa.
B7-H3 Monoclonal antibody specifically detects B7-H3 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
B7-H3 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q5ZPR3 | |
CD276 | |
Purified recombinant fragment of human CD276 (A.A. 30-130) expressed in E. Coli. | |
RUO | |
80381 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
6A1 | |
Unconjugated | |
CD276 | |
4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |