missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522248
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A375. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BTLA Polyclonal antibody specifically detects BTLA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
BTLA | |
Polyclonal | |
Unconjugated | |
BTLA | |
A630002H24; B and T lymphocyte associated; B and T lymphocyte associated protein; B and T lymphocyte attenuator; B- and T-lymphocyte attenuator; B- and T-lymphocyte-associated protein; BTLA; BTLA_HUMAN; BTLA1; CD272; CD272 antigen; FLJ16065; MGC129743 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q7Z6A9 | |
BTLA | |
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 164 of CD272 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |