missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Monoclonal Antibody (LG.3A10), Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen MA517902
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17902 targets CD27/TNFRSF7 in FACS, IHC, and IHC (F) applications and shows reactivity with Mouse samples. The MA5-17902 immunogen is mCD27 tag expressing clones AR0524 and AR0530.
CD27 Monoclonal antibody specifically detects CD27 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
CD27 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P41272 | |
CD27 | |
MCD27 tag expressing clones AR0524 and AR0530 | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen) | |
LG.3A10 | |
Unconjugated | |
CD27 | |
CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
Armenian Hamster | |
Protein G | |
RUO | |
21940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |