missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528036
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD27 Polyclonal antibody specifically detects CD27 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD27 | |
Polyclonal | |
Unconjugated | |
CD27 | |
CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P26842 | |
CD27 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 225 of Human CD27 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |