missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIL-R2 (DR5) Monoclonal Antibody (DR5-01-1), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119759
256.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes an extracellular domain of TRAIL-R2 (DR5).
TRAIL-R2 (DR5) Monoclonal antibody specifically detects TRAIL-R2 (DR5) in Human samples. It is validated for Flow Cytometry.Specifications
TRAIL-R2 (DR5) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
O14763 | |
TNFRSF10B | |
Recombinant fusion protein of human IgG heavy chain and extracellular domain of DR5. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
DR5-01-1 | |
FITC | |
TNFRSF10B | |
apoptosis inducing protein TRICK2A/2B; apoptosis inducing receptor TRAIL-R2; CD262; cytotoxic TRAIL receptor-2; death domain containing receptor for TRAIL/Apo-2L; death receptor 5; Dr5; dr-5; Fas-like protein; KILLER; KILLER/DR5; KILLER/DR5 TRAIL death-inducing receptor; Ly98; MK; p53-regulated DNA damage-inducible cell death receptor(killer); sCD262; soluble CD262; TNF receptor superfamily member 10b; TNF-related apoptosis-inducing ligand receptor 2; TNFRSF10B; TRAIL receptor 2; TRAILR2; TRAIL-R2; TRICK2; TRICK2A; TRICK2B; TRICKB; tumor necrosis factor receptor superfamily member 10B; tumor necrosis factor receptor superfamily, member 10b; tumor necrosis factor receptor-like protein ZTNFR9; UNQ160/PRO186; ZTNFR9 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
8795 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |