missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD26 Monoclonal Antibody (BA5b), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110160
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD26 Monoclonal antibody specifically detects CD26 in Human samples. It is validated for Flow Cytometry.Specifications
CD26 | |
Monoclonal | |
PE | |
DPP4 | |
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
1803 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
BA5b | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P27487 | |
DPP4 | |
A human T cell clone | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |