missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Monoclonal Antibody (CD25-3G10), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135144
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD25 Monoclonal antibody specifically detects CD25 in Human, Mammalia, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
CD25 | |
Monoclonal | |
FITC | |
IL2RA | |
ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
Mouse | |
purified | |
RUO | |
16184, 25704, 3559 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
CD25-3G10 | |
PBS with BSA and 0.1% sodium azide | |
P01589, P01590, P26897 | |
IL2RA | |
CD-25 receptor a-chain. | |
500 μL | |
Primary | |
Human, Mammalia, Mouse, Rat | |
Antibody | |
IgG1 |