missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Monoclonal Antibody (MEM-140), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119233
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
CD25 | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
P01589 | |
IL2RA | |
PHA-activated peripheral blood leucocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgM |
Flow Cytometry, Immunoprecipitation | |
MEM-140 | |
Unconjugated | |
IL2RA | |
ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
Mouse | |
sequential chromatography | |
RUO | |
3559 | |
4° C, do not freeze | |
Liquid |