Learn More
CD25 Monoclonal Antibody (ICO-105), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17650
256.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-7650 detects CD25 from human samples. MA1-7650 has been successfully used in ELISA and Immunochemistry procedures. The MA1-7650 immunogen is activated T and B cells.
CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for ELISA, Immunocytochemistry.Specifications
CD25 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
P01589 | |
IL2RA | |
Activated T and B cells. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
ICO-105 | |
Unconjugated | |
IL2RA | |
ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
Mouse | |
Protein A | |
RUO | |
3559 | |
4°C short term, -80°C long term | |
Liquid |