missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHCE Monoclonal Antibody (BRIC69), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA190231
Code : LE
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RHCE Monoclonal antibody specifically detects RHCE in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
RHCE | |
Monoclonal | |
1 mg/mL | |
tris glycine with 0.09% sodium azide | |
P18577 | |
RHCE | |
Human erythrocytes | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
BRIC69 | |
Unconjugated | |
RHCE | |
(C)ces type 1 Rhesus blood group D antigen; blood group Rh(CE) polypeptide; blood group RhCcEe antigen; blood group RhCE polypeptide; CD240CE; RH; Rh blood group antigen Evans; Rh blood group C antigen; Rh blood group CcEe antigens; Rh blood group protein; Rh blood group, CcEe antigens; rh polypeptide 1; Rh polypeptide I; Rh30A; Rh4; RHC; RHCE; RhCE blood group antigens; RHCE blood group variant Crawford antigen Rh43; RHE; rhesus blood group antigen, RhC antigen; Rhesus blood group CE protein; Rhesus blood group E antigen; rhesus blood group little e antigen; Rhesus blood group Rhce antigen; rhesus C/E antigens; Rhesus system C and E polypeptides; RhIVb(J); RHIXB; RhPI; RhVI; RhVIII; silenced Rh blood group CcEe antigen; truncated RhCcEe antigen; truncated RHCE | |
Mouse | |
purified | |
RUO | |
6006 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |