Learn More
CD235a Monoclonal Antibody (YTH89.1), Biotin, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517698
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17698 targets CD235a/Glycophorin A in FACS and WB applications and shows reactivity with Human samples. The MA5-17698 immunogen is rat monoclonal antibodies raised against human CD235a.
CD235a Monoclonal antibody specifically detects CD235a in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD235a | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P02724 | |
GYPA | |
Rat monoclonal antibodies raised against human CD235a. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
YTH89.1 | |
Biotin | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Rat | |
Protein G | |
RUO | |
2993 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |