missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD235a Monoclonal Antibody (HIR2), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119708
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes the N-terminal portion of glycophorin A (and weakly of glycophorin B).
CD235a Monoclonal antibody specifically detects CD235a in Human samples. It is validated for Flow Cytometry.Specifications
CD235a | |
Monoclonal | |
FITC | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
2993 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HIR2 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P02724 | |
GYPA | |
Synthetic peptide corresponding to the N-terminal region of human HIR2. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |