missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23 Mouse anti-Human, PE, Clone: 138628, Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA523549
Code : NEW
Additional Details : Weight : 0.00010kg
Description
CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.Specifications
CD23 | |
Monoclonal | |
PE | |
FCER2 | |
CD23, CD23A, CLEC4J, FCE2, IGEBF, BLAST-2, C-type lectin domain family 4, member J, CD23 antigen, fc-epsilon-RII, immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, lymphocyte IgE receptor | |
Mouse | |
Protein A/G | |
RUO | |
2208 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
138628 | |
Buffered saline solution with BSA and 0.1% sodium azide | |
P06734 | |
FCER2 | |
Human CD23 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |